Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Media Format:Liquid
- Description:Sabouraud Dextrose Broth
- Size:1 L
- Cat. No.:101373-444
- Cap type:33-430 black screw cap
- Fill volume:1000 ml
- Supplier no.:B1325
Specifications
About this item
Sabouraud Dextrose Broth is a prepared medium contained in a one-liter bottle.
- Black Screw Cap
- One Liter Bottle
- Fill Volume: 1000 mL
- Prepared Medium
NEL produces a wide variety of agar-based media in many sizes and types of bottles.
Clients in the pharmaceutical, hospital, food and beverage, cosmetic, and educational research industries depend upon the 500 proprietary products and over 1300 classic media formulations the NEL creates.