Order Entry
United States
ContactUsLinkComponent
R2A Agar, Bottles, Northeast Lab Services
R2A Agar, Bottles, Northeast Lab Services
Catalog # 101373-690
CAS Number:  
R2A Agar, Bottles, Northeast Lab Services
Catalog # 101373-690
Supplier Number:  B4060
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Media Format:
    Liquid
  • Description:
    R2A Agar
  • Size:
    500 mL
  • Cat. No.:
    101373-690
  • Cap type:
    33-430 black screw cap
  • Fill volume:
    350 ml
  • Supplier no.:
    B4060

Specifications

About this item

R2A Agar has a fill volume of 350 mL contained in a clear glass bottle of 500 mL.

  • Clear Glass Construction
  • Black Screw Cap
  • Prepared Medium
  • Bottle: 500 mL
  • Fill Volume: 350 mL

NEL produces a wide variety of agar-based media in many sizes and types of bottles.

NEL manufactures over 500 proprietary products and over 1300 classic media formulations for clients in the pharmaceutical, hospital, food and beverage, cosmetic, and educational research industries.