Order Entry
United States
ContactUsLinkComponent
OFPBL Agar, Plates, Northeast Lab Services
OFPBL Agar, Plates, Northeast Lab Services
Catalog # 101374-060
CAS Number:  
OFPBL Agar, Plates, Northeast Lab Services
Catalog # 101374-060
Supplier Number:  P1497
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Media Format:
    Liquid
  • Description:
    OFPBL Agar
  • Size:
    15 mm x 100 mm
  • Cat. No.:
    101374-060
  • Fill volume:
    20 ml
  • Supplier no.:
    P1497

Specifications

About this item

This OFPBL Agar is contained in a standard Petri Dish.

  • Standard Petri Dish
  • Fill Volume: 20 mL
  • Prepared Medium
  • Size: 15 mm x 100 mm

The petri dish is 15 mm x 100 mm, and the slant's fill volume is 20 mL.

NEL's plated media operation utilizes carefully selected and validated ingredients for contaminant-free products with a high degree of consistency from batch to batch.

Certifications: ISO Class 5 / FS209E Class 100 Validated EC Rooms certified.