Order Entry
United States
ContactUsLinkComponent
Schaedler Agar, Bottles, Northeast Lab Services
Schaedler Agar, Bottles, Northeast Lab Services
Catalog # 101373-720
CAS Number:  
Schaedler Agar, Bottles, Northeast Lab Services
Catalog # 101373-720
Supplier Number:  B5395
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Media Format:
    Liquid
  • Description:
    Schaedler Agar
  • Size:
    1 L
  • Cat. No.:
    101373-720
  • Cap type:
    Screw cap
  • Fill volume:
    500 ml
  • Supplier no.:
    B5395

Specifications

About this item

Schaedler agar includes a convenient marking spot and polypro cap.

  • Polypro Screw Cap
  • Marking Spot
  • Prepared Media
  • Fill Volume: 500 mL

The one-liter bottle shows graduations and a screw cap.

NEL manufactures over 500 proprietary products and over 1300 classic media formulations for clients in the pharmaceutical, hospital, food and beverage, cosmetic, and educational research industries.

Certifications: Follows FDA, GMP, and USDA guidelines.