About this item
The innuPREP Plasmid Mini Kit allows researchers to isolate plasmid DNA from bacterial lysates quickly and easily.
- A combination kit for extracting high-copy and low-copy plasmid DNA, P1 constructs, etc.
- Final yields of up to 40µg of highly pure plasmid DNA
- Starting volumes of the bacterial suspensions to be used covers a large range (0.5-10ml)
- The eluted pDNA can be used immediately prior to sequencing
The innuPREP Plasmid Mini Kit allows researchers to isolate plasmid DNA from bacterial lysates quickly and easily. The method combines alkaline lysis with the process of binding plasmid DNA to a filter membrane once the chromosomal DNA and bacterial proteins have been precipitated. The bound plasmid DNA is then washed and eluted by adding a low salt buffer. The resulting isolated plasmid DNA can then be used immediately in a variety of additional downstream applications. The new method allows scientists to isolate plasmid DNA from 0.5-10ml of starting material. The use of a novel binding membrane makes it possible to isolate up to 40µg of pDNA.
Specifications
Starting material: Bacterial suspensions; Isolation of high-copy plasmids: 0.5-5ml; Isolation of low-copy plasmid DNA, P1 constructions, etc: > 5-10ml
Column binding capacity: approx. 40µg pDNA
Average purity: 1.8 – 2.0
Extraction time: Approx. 16 minutes
Typical yield from 2 ml starting material (high-copy plasmid): 6-20µg
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Method / Format:
- Refine target molecule:
- Sample type:
- Target molecule:
- Environmentally Preferable: