ContactUsLinkComponent
3M™ Heavy Duty Duct Tape 3939, 3M Company
  89235-842
 :  3M
3M™ Heavy Duty Duct Tape 3939, 3M Company
  89235-842
 :  3M
 :  7000028933
  • Width:
    1.88"
  • Description:
    3M™ Heavy Duty Duct Tape 3939 has the tensile strength and adhesion for a host of other industrial applications.
  • Cat. No.:
    89235-842
  • Supplier no.:
    00051131069756
3M™ Heavy Duty Duct Tape 3939 is constructed of a water-resistant polyethylene laminated to cloth reinforcement and backed by an aggressive natural-rubber adhesive; the tape is high-tack and conformable.
  • High tensile strength for a secure hold under stress
  • Thick rubber adhesive bonds to most rough and smooth surfaces for a tight seal
  • Silver polyethylene surface resists wear, abrasion, and water
  • Good choice for hanging polyethylene drapes for containment activities
Our tape tears easily and cleanly—horizontally or vertically—without slivering
3M™ Heavy Duty Duct Tape 3939 can be used in temperatures up to 200 degrees F. It has been burn tested according to the methods UL 723, ASTM E-84, NFPA 255, UBC 8-1. Results were Flame Spread Index 0, Smoke Developed Index 30.
Duct tape (sometimes mistakenly referred to as “duck tape”) is a toolbox mainstay, used in maintenance, repair and construction operations. The term “duct tape” comes from the silver tape’s original use in heating, ventilation and air conditioning (HVAC) applications, including duct sealing. Today it performs wherever patching, bundling, reinforcing, tabbing, splicing insulation, capping pipe and conduit ends, labeling, temporary repair, hanging polyethylene drapes, or protecting against light sandblast overspray is needed. Duct tape is used in residential, industrial and commercial environments and by members of construction, electrical, industrial, marine, and military, abatement, trucking and automotive markets.
Industrial applications include hanging polyethylene drapes for containment activities, masking and sandblast overspray protection. It is also used in the convention industry for holding and seaming carpet padding, taping down cords and hinging poster-board panels.