About this item
The innuPREP Forensic Kit is a sophisticated tool optimized specifically for isolating genomic DNA from tiny samples and from highly contaminated forensic specimens.
- Optimal DNA extraction from very small forensic or highly contaminated samples
- Ready-to-use genomic DNA for immediate downstream application
- Large range of starting materials have been tested with positive results
- Certified for in-vitro diagnostic applications (CE-IVD)
The innuPREP Forensic Kit is a sophisticated tool optimized specifically for isolating genomic DNA from tiny samples and from highly contaminated forensic specimens. Extraction has been successfully tested for a large number of starting materials, such as blood, traces of blood, hair, hair roots, beard stubble, finger nails, postage stamps, cigarette butts, chewing gum, traces of sperm, swab samples, and finger prints on a variety of surfaces. The novel extraction chemistry also makes it possible to recover severely degraded nucleic acids. In addition, CE-IVD certification makes the innuPREP Forensic Kit suitable for use in in-vitro diagnostic applications.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Method / Format:
- Refine target molecule:
- Sample type:
- Target molecule:
- Environmentally Preferable: