Order Entry
United States
ContactUsLinkComponent
 
FEP Tubing, Pickering Laboratories
  100359-080
FEP Tubing, Pickering Laboratories
  100359-080
 :  2102-0463

 

  • Tubing ext.Ø:
    0.063"
  • Tubing int.Ø:
    ⅛"
  • Description:
    FEP Tube, Clear
  • Length:
    90 cm
  • Cat. No.:
    100359-080
  • Supplier no.:
    2102-0463

 

 

Clear tubing for one and two pump system.