Order Entry
United States
ContactUsLinkComponent
 
Anti-Mouse IgG Antibody (AP (Alkaline Phosphatase))
  PAS3721
undefined
Anti-Mouse IgG Antibody (AP (Alkaline Phosphatase))
  PAS3721
 :  S3721
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G
  • Antigen Symbol:
    IgG
  • Conjugation:
    AP (Alkaline Phosphatase)
  • Reactivity:
    Mouse
  • Size:
    100 μl
  • Storage Temperature:
    2 °C to 10 °C
  • Cat. No.:
    PAS3721
  • Supplier no.:
    S3721

 

 

Polyclonal secondary antibody raised in goat, immunoaffinity-purified using corresponding immobilized antigens and conjugated to alkaline phosphatase (AP) enzyme. The mouse antibodies react with all IgG subclasses.

  • Secondary Antibody That Binds to Both Heavy and Light Chains for All IgG Subclasses
  • Extensively used and published in numerous research articles
  • Antibody is ready-to-use

Polyclonal secondary antibody raised in goat, immunoaffinity-purified using corresponding immobilized antigens and conjugated to alkaline phosphatase (AP) enzyme. The product (unless otherwise noted) is supplied as a 1mg/ml solution. The Anti-Mouse IgG (H+L), AP Conjugate antibody binds to both heavy and light chains for all IgG subclasses. As with all antibodies, in certain applications some species-dependent antigen-dependent cross-reactivity may be observed. A starting working dilution of 1:2,500 is suggested for most Western blot, dot blot and ELISA applications. The optimum concentration of secondary antibody depends on the application and will need to be empirically determined.

Type:
Antigen: Halotag
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: