Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
DMH-1
DMH-1
DMH-1
:
Avantor
DMH-1
:
Avantor
:
1206711-16-1
:
:
C₂₄H₂₀N₄O
:
MFCD18384963
:
380.44 g/mol
:
3316
9542.pdf
ghs_9542.pdf
Frequently Bought Together
Clorox® Concentrated Germicidal Bleach
: Janitorial Supplies
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
: Kimberly-Clark
VWR® Powder-Free Nitrile Examination Gloves
: VWR International
VWR® Lab Markers
: VWR International
VWR® Broken Glass Disposal Cartons
: VWR International
VWR® Disposable Serological Pipettes
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Nitrile Examination Gloves, Black
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
: VWR International
PURPLE NITRILE™ and PURPLE NITRILE-XTRA™ Examination Gloves, Kimberly-Clark
: Ansell Healthcare
VWR® Disposable Petri Dishes
: VWR International