Adhesive Vinyl GHS OSHA Labels 'Calcium Hypochlorite', HCL Labels
Supplier: HCL Label
Certificates
About this item
These ready-made adhesive vinyl labels comply with the updated OSHA HazCom 'secondary container' labeling requirements.
- Comply with labeling requirements
- Chemical resistant lamination
- Two-year shelf/outdoor life
These labels are compliant with OSHA's adoption of the UN Globally Harmonized System for the Classification and Labeling of Chemicals (GHS).
Certifications: RoHS compliant.
Frequently Bought Together