ContactUsLinkComponent
Petroleum Sorbent Spill Response Pack, 3M Company
  67101-092
 :  3M
Petroleum Sorbent Spill Response Pack, 3M Company
  67101-092
 :  3M
 :  7000051852
  • Color:
    White
  • Description:
    Petroleum Sorbent Spill Response Pack
  • Size:
    4 Gallons (US)/Pack
    12 Gallons (US)/Case
  • Cat. No.:
    67101-092
  • Supplier no.:
    SRP-PETRO
This petroleum sorbent spill response pack is designed to sorb and contain small petroleum spills and leaks fast.
  • For use in oil-only Sorbent applications
  • Designed to absorb and contain small spills/leaks
  • Absorbs up to four gallons of petroleum based liquids
  • Compact size is convenient for trucks, shipping departments and for areas of oil transfer
  • Kit contains three mini-booms and five pads
This petroleum sorbent spill response pack is designed to sorb and contain small petroleum spills and leaks fast. The 3M™ Petroleum Sorbent Spill Response Pack SRP-Petro will sorb hydrocarbon based liquids and repel aqueous based liquids.