Order Entry
United States
ContactUsLinkComponent
 
PAH Interferences Standard, Restek
Catalog # 75835-042
Supplier:  Restek
CAS Number:  
PAH Interferences Standard, Restek
Catalog # 75835-042
Supplier:  Restek
Supplier Number:  32472
CAS Number:  

Specifications

  • Concentration:
    In toluene
  • Description:
    PAH Interferences Standard
  • Size:
    1 ml/Ampule
  • Cat. No.:
    75835-042
  • Supplier no.:
    32472

Specifications

About this item

  • The only standard formulated to contain all major PAH interferences for priority PAHs
  • Pair with an Rxi-PAH column and Restek’s EU 15+1 PAH standard to optimize method conditions and ensure separation of interferences and target PAHs
  • Confirm the presence of common interferences by retention time comparisons
  • Certified reference material (CRM) meets strict ISO quality requirements, making it suitable for quantitative use as a calibration or fortification standard
  • PAH components are present in different concentrations so isomers can be distinguished by the difference in response factors
  • Low-volatility toluene solvent provides longer shelf life, which helps ensure more accurate results
  • Broad solvent miscibility—compatible with acetonitrile, the QuEChERS solvent
  • HPLC compatible when diluted in appropriate solvent and analyzed with Pinnacle II PAH columns or UHPLC compatible on the Pinnacle DB PAH column.