GC Multiresidue Pesticide Standard #6 (18 Compounds), Restek
Catalog # 75835-056
Supplier: Restek
CAS Number:
Specifications
- Concentration:100 µg/ml each in toluene
- Description:GC Multiresidue Pesticide Standard #6
- Size:1 ml/Ampule
- Cat. No.:75835-056
- Supplier no.:32568
Specifications
About this item
Accurately identify and quantify pesticide residues by GC-MS/MS in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea and dietary supplements.
- Quantitatively tested to confirm composition, detailed support documentation provided
- Synthetic pyrethroid compounds
Formulated and grouped for maximum long-term stability* and well-balanced chromatographic performance.
Optimized multiresidue pesticide method is offered free of charge and downloadable XLS file includes conditions and transition tables.
Consists of 18 components, those are acrinathrin, anthraquinone, bifenthrin, bioallethrin, cyfluthrin, lambda-cyhalothrin, cypermethrin, deltamethrin, fenvalerate, flucythrinate, tau-fluvalinate, cis-permethrin, trans-permethrin, phenothrin, resmethrin, tefluthrin, tetramethrin and transfluthrin.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Ordering information: The Comprehensive 203-compound kit also available (cat. # 32562) that covers food safety lists by the FDA, USDA, and other global governmental agencies; other ampoules also sold separately.
* NOTE: When combining many compounds with different chemical functionalities, mix stability can be an issue. In formulating these standards, we extensively studied the 203 compounds involved, and then grouped them into as few mixes as possible while still ensuring maximum long-term stability and reliability. For quantitative analysis, we recommend analysing each mix separately to ensure accurate results for every compound.
* NOTE: When combining many compounds with different chemical functionalities, mix stability can be an issue. In formulating these standards, we extensively studied the 203 compounds involved, and then grouped them into as few mixes as possible while still ensuring maximum long-term stability and reliability. For quantitative analysis, we recommend analysing each mix separately to ensure accurate results for every compound.