ContactUsLinkComponent
GC Multiresidue Pesticide Kit, Restek
  75835-044
 :  Restek
 :  
GC Multiresidue Pesticide Kit, Restek
  75835-044
 :  Restek
 :  32562
 :  
  • Description:
    GC Multiresidue Pesticide Kit
  • Size:
    Contains 1 ml each of these mixtures
  • Cat. No.:
    75835-044
  • Supplier no.:
    32562
Accurately identify and quantify pesticide residues by GC-MS/MS in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea, and dietary supplements.
  • Formulated and grouped for maximum long-term stability
  • Quantitatively tested to confirm composition
  • Contains 1 ml each of these mixtures
GC multiresidue pesticide kit consist of GC multiresidue pesticide standard #1, GC multiresidue pesticide standard #2, GC multiresidue pesticide standard #3.
 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements. Comprehensive 203-compound kit covers food safety lists by the FDA, USDA.
 : * NOTE: When combining a large number of compounds with different chemical functionalities, mix stability can be an issue. In formulating these standards, we extensively studied the 203 compounds involved, then grouped them into as few mixes as possible while still ensuring maximum long-term stability and reliability. For quantitative analysis, we recommend analyzing each mix separately to ensure accurate results for every compound.