DHA PiONA Standard (133 Components), Restek
75834-588
: Restek
:
- Concentration:Neat
- Description:DHA PiONA Standard
- Size:0.15 ml in autosampler vial
- Cat. No.:75834-588
- Supplier no.:30730
This standard is produced for refineries performing Detailed Hydrocarbon Analysis (DHA) of crude oil feedstocks and fuels.
It contains 133 components.
It helps with calibrating complex hydrocarbon analyses and provides the greatest number of gravimetrically determined values for quantitative calibration.
: ASTM Methods D6729, D6730, and D6733 (Determination of Individual Components in Spark Ignition Engine Fuels)
: Quantity discounts not available.