EFSA PAH4 Standard, Restek
Catalog # 75835-038
Supplier: Restek
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:1000 µg/ml each in toluene
- Description:EFSA PAH4 Standard
- Size:1 ml/Ampule
- Cat. No.:75835-038
- Supplier no.:32469
Specifications
About this item
- European Food Safety Authority (EFSA) PAH4 compounds prepared in toluene solvent
- High 1,000 µg/mL concentration—dilute as needed to economically prepare custom mixes
- Low volatility—longer shelf life and more accurate results.
- High miscibility—compatible with QuEChERS solvents
- Certified reference material (CRM) meets strict ISO quality requirements
- Pair with Rxi-PAH GC column for easy separation of EFSA PAH4 compounds from common interferences in food and environmental samples: www.restek.com/rxi-pah