LC Multiresidue Pesticide Standard #8 (Hydramethylnon), Restek
Catalog # 75834-898
Supplier: Restek
CAS Number:
Specifications
- Concentration:100 µg/ml each in acetonitrile
- CAS No.:67485-29-4
- Description:Hydramethylnon
- Size:1 ml/Ampule
- Formula:C₂₅H₂₄F₆N₄
- Cat. No.:75834-898
- Supplier no.:31979
Specifications
About this item
Accurately detect and quantify pesticides of global food safety concern in a wide range of fruits, vegetables, and other commodities by LC-MS/MS.
- Quantitatively tested to confirm composition, detailed support documentation provided
- Organonitrogen compounds
Formulated and grouped for maximum long-term stability* and well-balanced chromatographic performance.
Optimized multiresidue pesticide method is offered free of charge and downloadable XLS file includes conditions and transition tables.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Ordering information: The complete 204-compound kit also available (cat. # 31971) that covers many LC-determined pesticides listed by government agencies, other ampoules also sold separately.
* NOTE: When combining many compounds with different chemical functionalities, mix stability can be an issue. In formulating these standards, we extensively studied the 204 compounds involved, and then grouped them into as few mixes as possible while still ensuring maximum long-term stability and reliability. For quantitative analysis, we recommend analyzing each mix separately to ensure accurate results for every compound.
* NOTE: When combining many compounds with different chemical functionalities, mix stability can be an issue. In formulating these standards, we extensively studied the 204 compounds involved, and then grouped them into as few mixes as possible while still ensuring maximum long-term stability and reliability. For quantitative analysis, we recommend analyzing each mix separately to ensure accurate results for every compound.