GC-FID Test Mix (3 Components), Restek
Catalog # 75835-354
Supplier: Restek
CAS Number:
Specifications
- Concentration:20 µg/ml each in hexane
- Description:GC-FID Test Mix
- Size:1 ml/Ampule
- Cat. No.:75835-354
- Supplier no.:35108
Specifications
About this item
The mix consists of (C12) n-dodecane, (C14) n-tetradecane and (C16) n-hexadecane.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.