ContactUsLinkComponent
EP 2.4.22 Composition of Fatty Acids by GC Mix 2 (5 Components), Restek
EP 2.4.22 Composition of Fatty Acids by GC Mix 2 (5 Components), Restek
  75835-352
 :  Restek
 :  
EP 2.4.22 Composition of Fatty Acids by GC Mix 2 (5 Components), Restek
  75835-352
 :  Restek
 :  35101
 :  
  • Description:
    EP 2.4.22 Composition of Fatty Acids by GC Mix 2
  • Size:
    100 mg total
  • Cat. No.:
    75835-352
  • Supplier no.:
    35101
The composition of fatty acids by GC mix 2 consists of five components they are, (C6:0) methyl caproate 10%, (C8:0) methyl caprylate, 10%, (C10:0) methyl decanoate 20%, (C12:0) methyl dodecanoate, 20% and (C14:0) methyl myristate 40%.