EP 2.4.22 Composition of Fatty Acids by GC Mix 1 (6 Components), Restek
Catalog # 75835-350
Supplier: Restek
CAS Number:
Specifications
- Concentration:100 mg total
- Description:EP 2.4.22 Composition of Fatty Acids by GC Mix 1
- Size:100 mg total
- Cat. No.:75835-350
- Supplier no.:35100
Specifications
About this item
The composition of fatty acids by GC mix 1 consists of 6 components, they are (C20:0) methyl arachidate 40%, (C12:0) methyl dodecanoate 5%, (C14:0) methyl myristate 5%, (C18:1 [cis9]) methyl oleate 20%, (C16:0) methyl palmitate 10% and (C18:0) methyl stearate 20%.