2-Propanol, Restek
Catalog # 75835-586
Supplier: Restek
CAS Number:
Specifications
- Concentration:50000 µg/ml in water
- CAS No.:67-63-0
- Description:2-Propanol
- Size:1 ml/Ampule
- Formula:C₃H₈O
- Description:2-Propanol
- Cat. No.:75835-586
- Supplier no.:30473
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.