Residual Solvents - Class 1 (5 Components), Restek
Catalog # 75835-462
Supplier: Restek
CAS Number:
Specifications
- Concentration:In dimethyl sulfoxide
- Description:Residual Solvents - Class 1
- Size:1 ml/Ampule
- Cat. No.:75835-462
- Supplier no.:36279
Specifications
About this item
The solvents consists of five components, they are benzene (10 mg/ml), carbon tetrachloride (20 mg/ml), 1,2-dichloroethane 25 mg/ml, 1,1-dichloroethene (40 mg/ml), and 1,1,1-trichloroethane (50 mg/ml).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.