Residual Solvents Class 2 - Mix A (15 components), Restek
Catalog # 75835-454
Supplier: Restek
CAS Number:
Specifications
- Concentration:In dimethyl sulfoxide
- Description:Residual Solvents Class 2 - Mix A
- Size:1 ml/Ampule
- Cat. No.:75835-454
- Supplier no.:36271
Specifications
About this item
These mixtures reflect the changes made in USP30/NF25 effective July 1, 2008.
Contains: Acetonitrile, 2,05 mg/ml, chlorobenzene, 1,80 mg/ml, cyclohexane , 19,40 mg/ml, cis-1,2-dichloroethene, 4,70 mg/ml, trans-1,2-dichloroethene, 4,70 mg/ml, 1,4-dioxane, 1,90 mg/ml, ethylbenzene, 1,84 mg/ml, methanol, 15,00 mg/ml, methylcyclohexane, 5,90 mg/ml, methylene chloride (dichloromethane), 3,00 mg/ml, tetrahydrofuran, 3,45 mg/ml, toluene, 4,45 mg/ml, m-xylene, 6,51 mg/ml, o-xylene, 0,98 mg/ml, p-xylene, 1,52 mg/ml.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.