PAH Supplement Mix for Method 8100 (8 Components), Restek
Catalog # 75835-810
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 µg/ml each in methylene chloride
- Description:PAH Supplement Mix for Method 8100
- Size:1 ml/Ampule
- Cat. No.:75835-810
- Supplier no.:31857
Specifications
About this item
The mix consists of benzo(j)fluoranthene, dibenz(a,h)acridine, dibenz(a,j)acridine, 7H-dibenzo(c,g)carbazole, dibenzo(a,e)pyrene, dibenzo(a,h)pyrene, dibenzo(a,i)pyrene, 3-methylcholanthrene.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.