1,4-Dioxane-d8, Restek
Catalog # 75834-504
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml in P and T methanol
- CAS No.:17647-74-4
- Description:1,4-Dioxane-d8
- Size:1 ml/Ampule
- Formula:C₄H₈O₂
- Description:1,4-Dioxane-d8
- Cat. No.:75834-504
- Supplier no.:30614