Order Entry
United States
ContactUsLinkComponent
Method 551.1 Pesticide/Herbicide Mix (16 Components), Restek
Method 551.1 Pesticide/Herbicide Mix (16 Components), Restek
Catalog # 75834-998
Supplier:  Restek
CAS Number:  
Method 551.1 Pesticide/Herbicide Mix (16 Components), Restek
Catalog # 75834-998
Supplier:  Restek
Supplier Number:  32438
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml each in acetone
  • Description:
    Method 551.1 Pesticide/Herbicide Mix
  • Size:
    1 ml/Ampule
  • Cat. No.:
    75834-998
  • Supplier no.:
    32438

Specifications

About this item

Contains 16 components, those are alachlor, atrazine, bromacil, cyanazine , endrin, endrin aldehyde, endrin ketone, γ-bhc, heptachlor, heptachlor epoxide, hexachlorobenzene, hexachlorocyclopentadiene, methoxychlor, metolachlor, simazine and trifluralin.

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.