Order Entry
United States
ContactUsLinkComponent
Quality Control Standard Mixture, Revised (2 Ccomponents), Restek
Quality Control Standard Mixture, Revised (2 Ccomponents), Restek
Catalog # 75835-736
Supplier:  Restek
CAS Number:  
Quality Control Standard Mixture, Revised (2 Ccomponents), Restek
Catalog # 75835-736
Supplier:  Restek
Supplier Number:  31641
CAS Number:  

Specifications

  • Concentration:
    500 µg/ml each in acetone (1 ml is enough mix to spike one 900 ml quality control sample), total hydrocarbon concentration is 1000 µg/ml
  • Description:
    Quality Control Standard Mixture, Revised
  • Size:
    1 ml/Ampule (1 ml is enough mix to spike one 900 ml quality control sample)
  • Cat. No.:
    75835-736
  • Supplier no.:
    31641

Specifications

About this item

Determination of hydrocarbon oil index - applicable to drinking, surface, waste, and treated water.

  • Updated reference materials for GC analysis of TPH in water
  • Total hydrocarbon concentration is 1000 μg/ml

The mix consists of diesel #2 (additive-free), motor oil (additive-free bp 325 - 460 or C18 - C32 retention time range).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.