Method 525.2 GC-MS Performance Check Mix, (3 components), Restek
Catalog # 75834-794
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 µg/ml each in acetone
- Description:Method 525.2 GC-MS Performance Check Mix
- Size:1 ml/Ampule
- Cat. No.:75834-794
- Supplier no.:31827
Specifications
About this item
This consists of 4,4'-DDT, DFTPP (decafl uorotriphenylphosphine) and endrin.