For use in oil-only Sorbent applications.
- Will 'sorb' hydrocarbon based liquids
- Mini-booms are typically used because of their tube-like shape to encircle spills and contain leaks
- Repels aqueous based liquids (hydrophobic)
Petroleum sorbent booms with scrim encased tubes of particulate sorbent material secured by a UV resistant netting
Petroleum sorbent mini-booms are a selective sorbent which will "sorb" hydrocarbon-based liquids (oleophilic), repel aqueous-based liquids (hydrophobic). Mini-booms are typically used because of their tube-like shape to encircle spills and contain leaks.
Petroleum sorbent pillows are designed with a scrim that encases particulate sorbent material. Pillows are used in areas where more capacity is desired than can easily be achieved using a sorbent pad.
High-capacity petroleum sorbent pads have a scrim on one side to increase strength and help eliminate linting. Conveniently sized pads allow the user to quickly spread sorbent over and in the path of a spill.
The petroleum sorbent drum cover is designed for a 55 gallon drum to catch drips and spills that occur during dispensing of all types of hydrocarbon materials used in daily operations. Will 'sorb' hydrocarbon based liquids, repel aqueous based liquids.
- Catalog No:
- 67100-826
- 67101-246
- 67101-224
- 67101-242
- 67101-244
- 67101-250
- 67101-230
- 67101-166
- 67101-168
- 67101-252
- 67100-882
- 67101-172
- 67101-174
- 67101-176
- Type:
- Hydrocarbon Oils - Fuel/Diesel/Motor; Mineral Oils
- Hydrocarbon Oils - Fuel/Diesel/Motor; Mineral Oils
- Hydrocarbon Oils - Fuel/Diesel/Motor; Mineral Oils
- Hydrocarbon Oils - Fuel/Diesel/Motor; Mineral Oils
- Hydrocarbon Oils - Fuel/Diesel/Motor; Mineral Oils
- Hydrocarbon Oils - Fuel/Diesel/Motor; Mineral Oils
- Hydrocarbon Oils - Fuel/Diesel/Motor; Mineral Oils
- Hydrocarbon Oils - Fuel/Diesel/Motor; Mineral Oils
- Mineral Oils; Hydrocarbon Oils - Fuel/Diesel/Motor
- Hydrocarbon Oils - Fuel/Diesel/Motor; Mineral Oils
- Hydrocarbon Oils - Fuel/Diesel/Motor; Mineral Oils
—
—
—