Pesticide Surrogate Mix (2 Components), Restek
Catalog # 75835-012
Supplier: Restek
CAS Number:
Specifications
- Concentration:In acetone 1 ml/ampule
- Matrix:Acetone
- Description:Pesticide Surrogate Mix
- Size:1 ml/Ampule
- Cat. No.:75835-012
- Supplier no.:32453
Specifications
About this item
Contains two components, those are decachlorobiphenyl and 2,4,5,6-tetrachloro-m-xylene.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.