Canadian Drinking Water Triazine Herbicides Mix (7 Components), Restek
Catalog # 75835-820
Supplier: Restek
CAS Number:
Specifications
- Concentration:500 µg/ml each
- Matrix:Acetone
- Description:Canadian Drinking Water Triazine Herbicides Mix
- Size:1 ml/Ampule
- Cat. No.:75835-820
- Supplier no.:31864
Specifications
About this item
The mix consists of alachlor, atrazine, cyanazine (bladex), metolachlor, metribuzin, prometryne, simazine.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.