To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:In methylene chloride
- Matrix:Methylene chloride
- Description:Internal Standard Mix, EPA 528
- Size:1 ml/Ampule
- Cat. No.:75834-752
- Supplier no.:31696
Specifications
About this item
This mix has two components: 3-Nitro-o-xylene (1,000 μg/ml) and 2,3,4,5-Tetrachlorophenol (2,000 μg/ml)
- Reference materials for U.S. EPA Method 528
- Fortification solution formulated based on MS sensitivity to each analyte.
EPA method 528 is a solid phase extraction (SPE)/gas chromatography/mass spectrometry screening method developed to measure 12 environmentally important phenols in drinking water and determine whether to regulate these contaminants in order to protect public health.
Method 528 detection limits for phenols range from 0,02 to 0,58 µg/l, concentrations below those needed for drinking water monitoring, based on current health effect information.
Developed a full set of standards for Method 528—calibration standard, internal standard, analyte fortification solution, surrogate standard, and GC/MS tune check solution. MS sensitivity to four of the phenols is significantly less than for the others, so we incorporate these at a higher concentration, 500µg/mL, in the fortification solution. Some of the substituted phenols exist as isomers that have similar mass spectra, but can be differentiated by retention time. The only coelutions are deuterated surrogates, which do not share common ions with the target analytes.
Shelf life 6 to 96 months.