Orders LinkContactUsLinkComponent
BOD Bottles
BOD Bottles
BOD Bottles
Certificates
BOD Bottles
2 Options Available Below
Product Details & Documents
For biochemical oxygen demand testing.
  • Glass bottles
  • Bottles are unnumbered
  • Feature penny head stopper
BOD Bottles are designed for incubating diluted samples of clean water, waste water, effluent water, and polluted water during the five-day biochemical oxygen demand (BOD) test.
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • 60 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $16.30
    $16.30
    Log in for availability
    Pack of 36
    Pack of 36 - $489.30
    $489.30
    Each
       $16.30
    Pack of 36
       $489.30
    Supplier Number: BOD060
  • 300 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $20.70
    $20.70
    Log in for availability
    Pack of 24
    Pack of 24 - $449.20
    $449.20
    Each
       $20.70
    Pack of 24
       $449.20
    Supplier Number: BOD300