Order Entry
United States
ContactUsLinkComponent
WA EPH Fractionation Check Mix (22 Components), Restek
WA EPH Fractionation Check Mix (22 Components), Restek
Catalog # 75835-684
Supplier:  Restek
CAS Number:  
WA EPH Fractionation Check Mix (22 Components), Restek
Catalog # 75835-684
Supplier:  Restek
Supplier Number:  31491
CAS Number:  

Specifications

  • Concentration:
    25 µg/ml each in hexane
  • Description:
    WA EPH Fractionation Check Mix
  • Size:
    1 ml/Ampule
  • Cat. No.:
    75835-684
  • Supplier no.:
    31491

Specifications

About this item

The mix consists of, (C8) n-octane, (C10) n-decane, (C12) n-dodecane, (C16) n-hexadecane, (C21) n-heneicosane, (C34) n-tetratriacontane, acenaphthene, acenaphthylene, anthracene, benz(a)anthracene, benzo(a)pyrene, benzo(b)fluoranthene, benzo(k)fluoranthene, benzo(ghi)perylene, chrysene, dibenz(a,h)anthracene, fluoranthene, fluorene, indeno(1,2,3-cd)pyrene, naphthalene, phenanthrene, pyrene.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.