Order Entry
United States
Orders LinkContactUsLinkComponent
PstI
PstI
Certificates
PstI

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

C TGCA/G G\ACGT C

  • Capable of digesting DNA in 15 minutes or less
  • Active and capable of digestion directly in GoTaq(R) Green Master Mix
  • Blue/white cloning qualified, provides a higher level of quality control for enzymes used in cloning applications

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • PAR6111
  • PAR6115
  • Enzyme Name:
  • PstI
  • PstI
  • Buffer supplied with enzyme:
  • Buffer H
  • Buffer H
  • Buffer activity:
  • % Activity (37°C) in 4 CORE® Buffers A: 10…25% – B: 50…75% – C: 50…75% – D: 50…75%; MULTI-CORE™ Buffer 25…50%
  • % Activity (37°C) in 4 CORE® Buffers A: 10…25% – B: 50…75% – C: 50…75% – D: 50…75%; MULTI-CORE™ Buffer 25…50%
  • Enzyme assay temperature:
  • 37°C
  • 37°C
  • Storage Conditions:
  • –30...–10 °C
  • –30...–10 °C

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • PstI
    3000 U
    Log in for availability
    Log in for availability
    Each
    Each - $50.55
    $50.55
    Each
       $50.55
    Minimum order quantity is 1

    Supplier Number: R6111
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    PAR6111
    Enzyme Name
    PstI
    Buffer supplied with enzyme
    Buffer H
    Buffer activity
    % Activity (37°C) in 4 CORE® Buffers A: 10…25% – B: 50…75% – C: 50…75% – D: 50…75%; MULTI-CORE™ Buffer 25…50%
    Enzyme assay temperature
    37°C
    Storage Conditions
    –30...–10 °C
  • PstI
    15000 U
    Log in for availability
    Log in for availability
    Each
    Each - $182.99
    $182.99
    Each
       $182.99
    Minimum order quantity is 1

    Supplier Number: R6115
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    PAR6115
    Enzyme Name
    PstI
    Buffer supplied with enzyme
    Buffer H
    Buffer activity
    % Activity (37°C) in 4 CORE® Buffers A: 10…25% – B: 50…75% – C: 50…75% – D: 50…75%; MULTI-CORE™ Buffer 25…50%
    Enzyme assay temperature
    37°C
    Storage Conditions
    –30...–10 °C