Order Entry
United States
ContactUsLinkComponent
Rsa I, Restriction Enzymes, Promega
Rsa I, Restriction Enzymes, Promega
Catalog # PAR6371
CAS Number:  
undefined
Rsa I, Restriction Enzymes, Promega
Catalog # PAR6371
Supplier Number:  R6371
CAS Number:  

Specifications

  • Description:
    Rsa I
  • Size:
    1000 U
  • Storage Conditions:
    –30...–10 °C
  • Enzyme Name:
    Rsa I
  • Buffer supplied with enzyme:
    Buffer C
  • Buffer activity:
    % Activity (37°C) in 4 CORE® Buffers A: 75…100% – B: 75…100% – C: 1 – D: < 10%; MULTI-CORE™ Buffer < 10%
  • Enzyme assay temperature:
    37°C
  • Cat. No.:
    PAR6371
  • Supplier no.:
    R6371

Specifications

About this item

GT/AC CA\TG

  • Capable of digesting DNA in 15 minutes or less
  • Available at high concentration, containing 5,000 units of RsaI at a concentration of 40-80u/microl