Order Entry
United States
Orders LinkContactUsLinkComponent
XhoI
XhoI
Certificates
XhoI

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

C/TCGA G G AGCT\C

  • Capable of digesting DNA in 15 minutes or less
  • Active and capable of digestion directly in GoTaq(R) Green Master Mix
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
  • Genome qualified to ensure optimal performance in genomic analysis applications

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • PAR6161
  • PAR6165
  • Enzyme Name:
  • XhoI
  • XhoI
  • Buffer supplied with enzyme:
  • Buffer D
  • Buffer D
  • Buffer activity:
  • % Activity (37°C) in 4 CORE® Buffers A: 25…50% – B: 75…100% – C: 75…100% – D: 1; MULTI-CORE™ Buffer 10…25%
  • % Activity (37°C) in 4 CORE® Buffers A: 25…50% – B: 75…100% – C: 75…100% – D: 1; MULTI-CORE™ Buffer 10…25%
  • Enzyme assay temperature:
  • 37°C
  • 37°C
  • Storage Conditions:
  • –30...–10 °C
  • –30...–10 °C

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • XhoI
    3000 U
    Log in for availability
    Log in for availability
    Each
    Each - $62.68
    $62.68
    Each
       $62.68
    Minimum order quantity is 1

    Supplier Number: R6161
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    PAR6161
    Enzyme Name
    XhoI
    Buffer supplied with enzyme
    Buffer D
    Buffer activity
    % Activity (37°C) in 4 CORE® Buffers A: 25…50% – B: 75…100% – C: 75…100% – D: 1; MULTI-CORE™ Buffer 10…25%
    Enzyme assay temperature
    37°C
    Storage Conditions
    –30...–10 °C
  • XhoI
    10000 U
    Log in for availability
    Log in for availability
    Each
    Each - $200.18
    $200.18
    Each
       $200.18
    Minimum order quantity is 1

    Supplier Number: R6165
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    PAR6165
    Enzyme Name
    XhoI
    Buffer supplied with enzyme
    Buffer D
    Buffer activity
    % Activity (37°C) in 4 CORE® Buffers A: 25…50% – B: 75…100% – C: 75…100% – D: 1; MULTI-CORE™ Buffer 10…25%
    Enzyme assay temperature
    37°C
    Storage Conditions
    –30...–10 °C