Order Entry
United States
ContactUsLinkComponent
Bgl I (Genome Qualified), Restriction Enzymes, Promega
Bgl I (Genome Qualified), Restriction Enzymes, Promega
Catalog # PAR6071
CAS Number:  
undefined
Bgl I (Genome Qualified), Restriction Enzymes, Promega
Catalog # PAR6071
Supplier Number:  R6071
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Bgl I (Genome Qualified)
  • Size:
    1000 U
  • Storage Conditions:
    –30...–10 °C
  • Enzyme Name:
    Bgl I (Genome Qualified)
  • Buffer supplied with enzyme:
    Buffer D
  • Buffer activity:
    % Activity (37°C) in 4 CORE® Buffers A: 10…25% – B: 25…% – C: 75…100% – D: 1; MULTI-CORE™ Buffer 1
  • Enzyme assay temperature:
    37°C
  • Cat. No.:
    PAR6071
  • Supplier no.:
    R6071

Specifications

About this item

GCCN NNN/NGGC CGGN\NNN NCCG

  • Genome qualified to ensure optimal performance in genomic analysis applications