Order Entry
United States
ContactUsLinkComponent
BamHI, Restriction Enzymes, Promega
BamHI, Restriction Enzymes, Promega
Certificates
undefined
BamHI, Restriction Enzymes, Promega
Certificates

Specifications

  • Storage Conditions:
    –30...–10 °C
  • Enzyme Name:
    BamHI
  • Buffer supplied with enzyme:
    Buffer E
  • Buffer activity:
    % Activity (37°C) in 4 CORE® Buffers A: 75…100% ** – B: 75…100% – C: 75…100% – D: 50…75%; MULTI-CORE™ Buffer 75…100%
  • Enzyme assay temperature:
    37°C

Specifications

Related Information

Frequently Bought Together