ContactUsLinkComponent
BBL™ Clostridium Difficile Selective Agar, BD
Catalog # 90006-588
Supplier:  Avantor
CAS Number:  
BBL™ Clostridium Difficile Selective Agar, BD
Catalog # 90006-588
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    BBL™ Clostridium Difficile Selective Agar
  • Size:
    10 x 100 mm; 15 x 100 mm
  • Environmentally Preferable:
  • Cat. No.:
    90006-588
  • Supplier no.:
    222228
Specifications
About this item
Clostridium difficile Selective Agar (CDSA) is recommended as a selective and differential medium for the primary isolation of Clostridium difficile from fecal specimens.
  • Amino acids are utilized to increase pH
  • Cefoxitin and cycloserine inhibit normal fecal flora
  • Agar is the solidifying agent
C. difficile colonies produce yellow fluorescence when viewed with long-wave UV light.