KBr Disc Starter Pack for Solids, Spectrum RX1 System, PerkinElmer
Supplier: PerkinElmer
Certificates
About this item
The complete pack contains an evacuable potassium bromide die for the preparation of 13mm diameter pellets of the highest quality and clarity, a pellet holder and adjustable mount for precise mounting of samples in your Spectrum RX, the easy-to-operate 15-tonne hydraulic press and 100g of research-grade KBr powder.
Ordering information: Starter pack is available with or without the hydraulic press.
Frequently Bought Together