ContactUsLinkComponent
BD BBL™ CHROMagar™ Staph aureus, BD Diagnostics
BD BBL™ CHROMagar™ Staph aureus, BD Diagnostics
Catalog # 90000-836
Supplier:  BD
CAS Number:  
undefined
BD BBL™ CHROMagar™ Staph aureus, BD Diagnostics
Catalog # 90000-836
Supplier:  BD
Supplier Number:  214982
CAS Number:  
Specifications
  • Description:
    BBL™ CHROMagar™ Staph aureus
  • Size:
    100 mm
  • Cat. No.:
    90000-836
  • Supplier no.:
    214982
Specifications
About this item
BBL™ CHROMagar™ Staph aureus is a selective medium for the isolation, enumeration and identification of Staphylococcus aureus.
  • Peptones supply essential nutrients
  • Additional selective agents inhibits gram-negative bacteria, yeast, and some gram-positive cocci
  • Chromogen mix consists of artificial substrates which release color when hydrolyzed
BBL™ CHROMagar™ Staph aureus is intended for the isolation, enumeration, and identification of S. aureus based on the formation of mauve-colored colonies. The addition of chromogenic substrates to the medium facilitates the differentiation of S. aureus from other organisms.