ContactUsLinkComponent
Desiccant Packs, Desco
Catalog # 89024-400
Supplier:  Desco
undefined
Desiccant Packs, Desco
Catalog # 89024-400
Supplier:  Desco
Supplier Number:  1/2KDES550
Specifications
  • Description:
    Desiccant Kraft Paper, ½ Unit, Pail of 550
  • Dimensions:
    6 x 8.9 cm (2.4 x 3.5")
  • Cat. No.:
    89024-400
  • Supplier no.:
    1/2KDES550
Specifications
About this item
Desiccant pouches are designed to absorb airborne moisture left inside packaging following vacuum sealing
These pouches capture moisture that is able to pass through the bag material, which helps to keep devices dry through unexpected shipping delays or extended storage time. Secured in a strong pouch of economical kraft paper.