LDH Cytotoxicity WST Assay, Enzo Life Sciences
Supplier: Enzo Life Sciences
Certificates
About this item
Colorimetric kit for the determination of cytotoxicity through the measurement of lactate dehydrogenase activity.
- Homogenous assay using living cells or non-homogenous assay protocol using cell culture supernatant
- Stable working solution
The LDH cytotoxicity WST assay is a colorimetric assay kit used to determine cytotoxicity by measuring lactate dehydrogenase activity released from damaged cells.
Frequently Bought Together