Order Entry
United States
ContactUsLinkComponent
Insulin ELISA Kit, Enzo Life Sciences
Insulin ELISA Kit, Enzo Life Sciences
Catalog # 75786-342
Supplier:  Enzo Life Sciences
CAS Number:  
Insulin ELISA Kit, Enzo Life Sciences
Catalog # 75786-342
Supplier:  Enzo Life Sciences
Supplier Number:  KIT141-0001
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Host:
  • Primary antibody reactivity:
    Pig
    Human
  • Target protein:
    Insulin
  • Description:
    Insulin ELISA Kit
  • Size:
    1 x 96 wells
  • Environmentally Preferable:
  • Sample Type:
    Serum, plasma, tissue culture media
  • Cross Reactivity:
    No cross-reactivity to similar family molecules (proinsulin).
  • Detection Range:
    15.6-500 pg/ml
  • Sensitivity:
    15.5 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    75786-342
  • Supplier no.:
    KIT141-0001

Specifications

About this item

Highly sensitive Insulin ELISA kit enabling detection of insulin in serum, plasma, tissue culture media in just 3 hours.

  • Highly sensitive measurement of insulin, detecting as little as 21.6 pg/ml (0.622 µIU/ml)
  • Negligible cross reactivity with proinsulin
  • High throughput format with results in 3 hours for up to 40 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis

For the quantitative determination of insulin in serum, plasma, and tissue culture media.

he Insulin ELISA kit is a colorimetric immunometric immunoassay kit with results in 3 hours.

Ordering information: Contains Microtiter plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Antibody Diluent, Conjugate, Substrate, and Stop Solution.