Order Entry
United States
ContactUsLinkComponent
ACTH ELISA Kit, Enzo Life Sciences
ACTH ELISA Kit, Enzo Life Sciences
Catalog # 75786-336
Supplier:  Enzo Life Sciences
CAS Number:  
ACTH ELISA Kit, Enzo Life Sciences
Catalog # 75786-336
Supplier:  Enzo Life Sciences
Supplier Number:  KIT138-0001
CAS Number:  

Specifications

  • Target protein:
    Adrenocorticotropic Hormone (ACTH)
  • Description:
    ACTH ELISA Kit
  • Size:
    1 x 96 wells
  • Sensitivity:
    0.46 pg/ml (0.5-165 pg/ml)
  • Cat. No.:
    75786-336
  • Supplier no.:
    KIT138-0001

Specifications

About this item

Highly sensitive ACTH ELISA kit enabling detection of ACTH in human plasma in 4.5 hours.

  • Sensitive measurement of ACTH, detecting as little as 0.46 pg/ml
  • Negligible reactivity to similar hormones
  • High throughput with results in 4.5 hours
  • Fully quantitative results that surpass semi-quantitative Western blot analysis

For the quantitative determination of ACTH in plasma in human samples.

The ACTH ELISA kit is a colorimetric immunoassay kit with results in 4.5 hours.