Vials, National Scientific™, Thermo Scientific
These vials are fully lot tested including HPLC and GC analysis for 15 critical parameters.
- Superior quality 1st hydrolytic class glass (Type 1) in clear and amber
- Fixed insert or conical base vials for limited volume sampling
Manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) glass.
: Vials are available in assembled and unassembled kits.
Frequently Bought Together