With our high purity albumins, researchers can formulate and customize their own specialized media for specific applications.
- Reduce shear in bioreactive systems
- Bind non-specific toxic compounds
- Serological diluent
- Molecular weight standard
- Tissue culture growth nutrient
Equitech-Bio offers a wide range of heat-shock and proprietary Cohn method processed animal serum albumins. With our high purity albumins, researchers can formulate and customize their own specialized media for specific applications. Our albumins are especially useful for creating defined serum-free or serum-reduced mixtures from basal media, reducing shear in bioreactor systems, and binding non-specific toxic compounds. Equitech-Bio offers albumins as a powder as well as a sterile solution. All of Equitech-Bio’s BSA is certified US origin.
Can be used as a serological diluent, as a molecular weight standard, and as a growth nutrient in tissue culture. Tested negative for Blue Tongue and Vesicular Stomatitis.
Ash content: ≤2.0%
Heavy metals: ≤20 ppm
Moisture: ≤5.0%
- Source:Serum
- Species:Bovine
- Storage Conditions:5 °C ±3 °C
- Form:Powder
- Endotoxin-free:Yes
- Protease-free:Y
- Protein Synonyms:BSA|Fraction V|Cohn Fraction V
- Protein/Peptide Name:Bovine Serum Albumin
- Solubility:≤0.15
- Color:Off-shade white
- Concentration:≥95%
- Formulation:Powder