Order Entry
United States
ContactUsLinkComponent
OQ Response Linearity Test Standard (6 components), Restek
OQ Response Linearity Test Standard (6 components), Restek
Catalog # 10058-058
Supplier:  Restek
CAS Number:  
undefined
OQ Response Linearity Test Standard (6 components), Restek
Catalog # 10058-058
Supplier:  Restek
Supplier Number:  33906
CAS Number:  

Specifications

  • Description:
    QO Response Linearity Test Standard
  • Size:
    1 ml/Ampule
  • Cat. No.:
    10058-058
  • Supplier no.:
    33906

Specifications

About this item

OQ response linearity test standard includes n-heptadecane (C17) 1.5 µg/mL, n-octadecane (C18) 10 µg/mL, n-nonadecane (C19) 2 µg/mL, n-eicosane (C20) 100 µg/mL, n-docosane (C22) 1000 µg/mL, n-tetracosane (C24) 10 000 µg/mL in isooctane, 1 mL/ampoule.